missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonicotinic Acid 99.0+%, TCI America™
$44.28 - $140.54
Chemical Identifiers
| CAS | 55-22-1 |
|---|---|
| Molecular Formula | C6H5NO2 |
| Molecular Weight (g/mol) | 123.111 |
| MDL Number | MFCD00006429 |
| InChI Key | TWBYWOBDOCUKOW-UHFFFAOYSA-N |
| Synonym | isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french |
| PubChem CID | 5922 |
| ChEBI | CHEBI:6032 |
| IUPAC Name | pyridine-4-carboxylic acid |
| SMILES | C1=CN=CC=C1C(=O)O |
Chemical Identifiers
| 55-22-1 | |
| 123.111 | |
| TWBYWOBDOCUKOW-UHFFFAOYSA-N | |
| 5922 | |
| pyridine-4-carboxylic acid |
| C6H5NO2 | |
| MFCD00006429 | |
| isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french | |
| CHEBI:6032 | |
| C1=CN=CC=C1C(=O)O |
Specifications
| 55-22-1 | |
| White | |
| C6H5NO2 | |
| 25 g | |
| TWBYWOBDOCUKOW-UHFFFAOYSA-N | |
| pyridine-4-carboxylic acid | |
| 5922 | |
| 123.11 | |
| Crystalline Powder |
| 315°C | |
| 3.0 to 4.0 (6g/L H2O 20°C) | |
| MFCD00006429 | |
| isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french | |
| C1=CN=CC=C1C(=O)O | |
| 123.111 | |
| CHEBI:6032 | |
| ≥99.0% (HPLC,T) | |
| Isonicotinic Acid |
Safety and Handling
EINECSNumber : (9)-0185
RTECSNumber : NS1103000
TSCA : Yes