missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinamide 99.0+%, TCI America™

Catalog No. I013525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I013525G 25 g
I0135500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I013525G Supplier TCI America Supplier No. I013525G
Only null left

Chemical Identifiers

CAS 1453-82-3
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00006432
InChI Key VFQXVTODMYMSMJ-UHFFFAOYSA-N
Synonym isonicotinamide, 4-pyridinecarboxamide, 4-carbamoylpyridine, isonicotinic acid amide, isonicotineamide, gamma-pyridinecarboxamide, pyridine-4-carboxylic acid amide, unii-4h3bh6yx9q, .gamma.-pyridinecarboxamide, 4h3bh6yx9q
PubChem CID 15074
ChEBI CHEBI:6031
IUPAC Name pyridine-4-carboxamide
SMILES C1=CN=CC=C1C(=O)N

Specifications

Melting Point 158°C
Color White
Quantity 25 g
Formula Weight 122.13
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinamide
RTECSNumber NR9500000
TSCA Yes