missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinamide, 99%

Catalog No. AAA1819018
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1819018 50 g
AAA1819022 100 g
AAA1819030 250 g
AAA1819036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1819018 Supplier Thermo Scientific Chemicals Supplier No. A1819018
Only null left

CAS: 1453-82-3 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1453-82-3
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00006432
InChI Key VFQXVTODMYMSMJ-UHFFFAOYSA-N
Synonym isonicotinamide, 4-pyridinecarboxamide, 4-carbamoylpyridine, isonicotinic acid amide, isonicotineamide, gamma-pyridinecarboxamide, pyridine-4-carboxylic acid amide, unii-4h3bh6yx9q, .gamma.-pyridinecarboxamide, 4h3bh6yx9q
PubChem CID 15074
ChEBI CHEBI:6031
IUPAC Name pyridine-4-carboxamide
SMILES C1=CN=CC=C1C(=O)N

Specifications

Melting Point 155°C to 158°C
Quantity 50 g
Beilstein 2173
Formula Weight 122.13
Percent Purity 99%
Chemical Name or Material Isonicotinamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-926-2
RTECSNumber NR9500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only