missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isomaltotriose 97.0+%, TCI America™
$163.28 - $759.44
Chemical Identifiers
| CAS | 3371-50-4 |
|---|---|
| Molecular Formula | C18H32O16 |
| Molecular Weight (g/mol) | 504.438 |
| MDL Number | MFCD00070623 |
| InChI Key | FBJQEBRMDXPWNX-IGSOQDMUSA-N |
| Synonym | isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol |
| PubChem CID | 133126476 |
| IUPAC Name | (3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol |
| SMILES | C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I0329100MG
|
TCI America
I0329100MG |
100 mg |
Each for $163.28
|
|
|||||
|
I03291G
|
TCI America
I03291G |
1 g |
Each for $759.44
|
|
|||||
Chemical Identifiers
| 3371-50-4 | |
| 504.438 | |
| FBJQEBRMDXPWNX-IGSOQDMUSA-N | |
| 133126476 | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O |
| C18H32O16 | |
| MFCD00070623 | |
| isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol | |
| (3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol |
Specifications
| 3371-50-4 | |
| C18H32O16 | |
| 100 mg | |
| FBJQEBRMDXPWNX-IGSOQDMUSA-N | |
| (3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol | |
| 133126476 | |
| ≥97.0% (HPLC) | |
| Isomaltotriose |
| White | |
| MFCD00070623 | |
| isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O | |
| 504.438 | |
| 504.44 | |
| Crystalline Powder |
Safety and Handling
TSCA : No