missing translation for 'onlineSavingsMsg'
Learn More

Isomaltotriose 97.0+%, TCI America™

Catalog No. I0329100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0329100MG 100 mg
I03291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0329100MG Supplier TCI America Supplier No. I0329100MG

Chemical Identifiers

CAS 3371-50-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00070623
InChI Key FBJQEBRMDXPWNX-IGSOQDMUSA-N
Synonym isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol
PubChem CID 133126476
IUPAC Name (3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 504.44
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Isomaltotriose