missing translation for 'onlineSavingsMsg'
Learn More

Isomaltose 97.0+%, TCI America™

Catalog No. p-7131286
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0231100MG 100 mg
I02311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0231100MG Supplier TCI America Supplier No. I0231100MG
Only null left

Chemical Identifiers

CAS 499-40-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00065373
InChI Key DLRVVLDZNNYCBX-MYBWVJKRSA-N
Synonym isomaltose
PubChem CID 133612057
IUPAC Name (2S,3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)O)O)O)O)O)O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 342.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isomaltose