missing translation for 'onlineSavingsMsg'
Learn More

Isoluminol 98.0+%, TCI America™

Catalog No. p-7118267
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53001G 1 g
A530010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53001G Supplier TCI America Supplier No. A53001G
Only null left

Chemical Identifiers

CAS 3682-14-2
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.16
MDL Number MFCD00010560
InChI Key HUDPLKWXRLNSPC-UHFFFAOYSA-N
Synonym 4-aminophthalhydrazide, isoluminol, 6-aminophthalazine-1,4-diol, chembl14882, 1,4-phthalazinedione, 6-amino-2,3-dihydro, 6-amino-2,3-dihydro-1,4-phthalazinedione, iso-luminol, 1enu, 4-aminophthaloylhydrazine, 4-amino-phthalsaeure-hydrazid
PubChem CID 95014
IUPAC Name 6-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione
SMILES NC1=CC=C2C(=O)NNC(=O)C2=C1

Specifications

Melting Point 300°C
Color White-Yellow
Quantity 1 g
Formula Weight 177.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoluminol [Chemiluminescence reagent]