missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Isolator Wrap™ Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 Contact Plates

Catalog No. R111820
Change view
Click to view available options
Quantity:
10/Pk.
Format:
55mm Contact Plate
Media Type:
Tryptic Soy Agar
Includes:
Lecithin, Polysorbate 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Includes
R111820 10/Pk. 55mm Contact Plate Tryptic Soy Agar Lecithin, Polysorbate 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R111820 Supplier Thermo Scientific™ Supplier No. NRE4782

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Obtain fast and accurate results with a contact plate designed for environmental monitoring.

Effortlessly sample your environment using Thermo Scientific™ Isolator Wrap™ Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 Contact Plates.

  • Isolator Wrap plates are for environmental sampling.
  • These products are manufactured in a cleanroom environment and validated sterile by gamma radiation.
  • Contact plates use GripFit™ technology, which ensures the lid remains in place during handling.
  • Plates are shrink-wrapped and contained in a protective outer barrier wrap, which is impermeable to isolator decontamination procedures but maintains the integrity of the medium inside.
  • An extra sterile bag is provided for transporting the plates to the laboratory for testing.
  • Not all products are available for sale in all territories.
  • Please inquire - Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Certifications/Compliance ISO 7/8 (Grade C and D)
Color Clear agar light to med amber
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Lecithin, Polysorbate 80
Form Prepared Medium
Format 55mm Contact Plate
Manufacturing Origin United States
Media Type Tryptic Soy Agar
Product Type Agar Plate
Sterility Sterile
Test Type Environmental Test
Fill Volume 15 mL
For Use With (Application) Environmental Monitoring
Includes Lecithin, Polysorbate 80
Packaging Type Flow Wrapped - Single Layer and Extra Sterile Bag
Quantity 10/Pk.
Diameter (Metric) 55 mm
Show More Show Less