missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Isolator Wrap Sterile Contact Sabouraud Dextrose 5.6 Agar Plates with Lecithin & Polysorbate 80 - Room Temperature

Catalog No. R111825RT
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R111825RT 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R111825RT Supplier Thermo Scientific™ Supplier No. R111825RT/DUP

Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate with Lecithin & Polysorbate 80 is used for the cultivation of yeasts and fungi.

  • Settling pink plates (single bag and double bag) are VHP resistant manufactured in a clean room environment. This provides the highest level of sterility assurance with no compromise of growth characteristics.
  • Each lot is tested to ensure consistent, reproducible results.
  • Storage Condition: 2 to 25°C.

Specifications

Content And Storage 20°C to 25°C, Room Temperature
Media Type Sabouraud Dextrose Agar
Product Type Contact Plate
For Use With (Equipment) RABS, Isolators and Cleanrooms
Quantity 10/Pk.
Diameter (Metric) 85 mm