missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Isolator Sterile Room Temperature TSA Standard Plate

Catalog No. R111816RT
Change view
Click to view available options
Format:
Mono plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R111816RT Mono plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R111816RT Supplier Thermo Scientific™ Supplier No. R111816RT

Sterile Tryptic Soy Agar standard plate meets or exceeds USP growth promotion requirements for use in Environmental monitoring and other sterile processes.

  • Isolator Sterile room temperature Tryptic Soy Agar plates are a general purpose growth media, which are complimentary to areas that do not have refrigeration near operational manufacturing.
  • Plates are compliant with USP and meet global standards for Lab use only microbiological testing.
  • They can be used in any ISO standard environnment.
  • Sufficient for sterile or non-sterile environments.
  • The plates meet the requirements for all areas of environmental sampling.

Specifications

Color Light amber
Content And Storage Agar 2°C to 25°C
Description Isolator settling plate
Format Mono plate
Manufacturing Origin United States
For Use With (Application) Environmental monitoring