Learn More
Poltex Isolation Gown Dispenser
Catalog No. p-200089277
Change view
missing translation for 'orderingAttributeHoverText'
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
Catalog No. Mounting
21377558 Magnetic Mount
21377559 VHB Tape
21377560 Wall Mount
3 options
Catalog No. 21377558
Supplier: Poltex GOWN1M
Only null left
Poltex Gown Dispenser holds a stack of isolation gowns or coveralls.
Hold a stack of isolation gowns or coveralls.
Durable and cleanable PETG construction.
Choose mounting method:
  • Wall mount (screws/anchors included)
  • Magnets (powerful magnets indcluded)
  • VHB Tape (very high bond tape included)
Specifications
Product Type Bulk Dispenser
Color Clear
Holds Isolation Gowns or Coveralls
Material PETG
Mounting Magnetic Mount
No. of Compartments 1
Product Style Open Top and Front
Sterility Non-sterile