missing translation for 'onlineSavingsMsg'
Learn More

Isohexane (mixture of isomeric branched chain Hexanes) 98.0+%, TCI America™

Catalog No. I013425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I013425ML 25 mL
I0134500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I013425ML Supplier TCI America Supplier No. I013425ML

Chemical Identifiers

CAS 73513-42-5
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009406
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
IUPAC Name 2-methylpentane
SMILES CCCC(C)C

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1208
Formula Weight 86.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material Isohexane (mixture of isomeric branched chain Hexanes)
EINECSNumber (2)-0006
TSCA No