missing translation for 'onlineSavingsMsg'
Learn More

Isoguvacine Hydrochloride 98.0+%, TCI America™

Catalog No. I087125MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I087125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I087125MG Supplier TCI America Supplier No. I087125MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68547-97-7
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
MDL Number MFCD00055192
InChI Key SUWREQRNTXCCBL-UHFFFAOYSA-N
Synonym isoguvacine hydrochloride, 1,2,3,6-tetrahydro-4-pyridinecarboxylic acid hydrochloride, 1,2,3,6-tetrahydropyridine-4-carboxylic acid hydrochloride, 4-pyridinecarboxylic acid, 1,2,3,6-tetrahydro-, hydrochloride, 1,2,3,6-tetrahydro-pyridine-4-carboxylic acid hydrochloride, 1,2,5,6,-tetrahydroisonicotinic acid hydrochloride, isoguvacine hcl, spectrum1502129, isoguvacine hydrochloride, solid
PubChem CID 155107
IUPAC Name 1,2,3,6-tetrahydropyridine-4-carboxylic acid;hydrochloride
SMILES C1CNCC=C1C(=O)O.Cl

Specifications

Melting Point 280°C
Color Yellow
Quantity 25 mg
Formula Weight 163.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Isoguvacine Hydrochloride
TSCA No
Recommended Storage Refrigerator