Learn More
Stoelting™ Isoflurane Vaporizers
Stoelting™ Isoflurane Vaporizers are used on the Gas Anesthesia Systems to ensure that the output concentration is not affected by flow rate, temperature variation, duration of use, liquid level, or back pressure fluctuation.
Supplier:  Stoelting™ 53903
 View more versions of this product
Catalog No. 10000929
Only null left
Description
Description
  • Specifically calibrated for use with LINK 7
  • Extended capacity
  • 10 year warranty contingent upon a 2 year service cycle
  • Larger capacity for liquid anesthetic means filling the vaporizer less often which, in turn, reduces exposure to anesthetic gases.
  • Available with either funnel fill or pin index/key fill systems.
Specifications
Chemical Identifiers
Neuroscience
250 mL
Specifications
Neuroscience
Link 7 only
250 mL
Standard funnel fill
Product Suggestions
Product Suggestions