Learn More
Beckman Coulter Canada Lp IsoFlow Sheath Fluid DFS Item
Supplier:  Beckman Coulter 8546859
This isotonic fluid is specifically formulated for use in flow cytometers. IsoFlow Sheath Fluid is carefully manufactured for low particule and fluorescence backgrounds to ensure superior signal to noise ratio measurements.
Catalog No. NC9455400
Only null left
Explore available promotions