missing translation for 'onlineSavingsMsg'
Learn More

Isoeugenol, predominantly trans, ≥99% (sum of cis - and trans -isomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111014790
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014790 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014790 Supplier Merck Emd Millipore Supplier No. 575221ML
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 5932-68-3
Linear Formula C10H12O2
MDL Number MFCD00009285
Type Analytical Standard
Quantity 1 mL
Formula Weight 164.20
Percent Purity ≥99% (sum of cis - and trans -isomers, GC)
Grade Analytical Standard