missing translation for 'onlineSavingsMsg'
Learn More

Isocytosine 98.0+%, TCI America™

Catalog No. I08145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08145G Supplier TCI America Supplier No. I08145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-53-2
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00057557 MFCD00023256
InChI Key XQCZBXHVTFVIFE-UHFFFAOYSA-N
Synonym isocytosine, 2-amino-4-hydroxypyrimidine, 2-aminopyrimidin-4-ol, 2-aminopyrimidin-4 1h-one, 2-amino-4-pyrimdinol, 2-amino-4-pyrimidone, 2-amino-4-oxypyrimidine, 2-aminouracil, 4-pyrimidinol, 2-amino, 4 1h-pyrimidinone, 2-amino
PubChem CID 66950
ChEBI CHEBI:55502
IUPAC Name 2-amino-3,4-dihydropyrimidin-4-one
SMILES NC1=NC=CC(=O)N1

Specifications

Melting Point 266°C
Color White
Quantity 5 g
Formula Weight 111.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isocytosine