missing translation for 'onlineSavingsMsg'
Learn More

Isocytosine, 99%

Catalog No. p-4089418
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC398320050 5 g Glass bottle
AC398320250 25 g Glass bottle
2 options

Catalog No. AC398320050

Supplier: Thermo Scientific Chemicals 398320050

Only null left

CAS: 108-53-2 | C4H5N3O | 111.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-53-2
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00057557 MFCD00023256
InChI Key XQCZBXHVTFVIFE-UHFFFAOYSA-N
Synonym isocytosine, 2-amino-4-hydroxypyrimidine, 2-aminopyrimidin-4-ol, 2-aminopyrimidin-4 1h-one, 2-amino-4-pyrimdinol, 2-amino-4-pyrimidone, 2-amino-4-oxypyrimidine, 2-aminouracil, 4-pyrimidinol, 2-amino, 4 1h-pyrimidinone, 2-amino
PubChem CID 66950
ChEBI CHEBI:55502
SMILES NC1=NC=CC(=O)N1

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information (50mg/ml in DMSO) Clear solution
IUPAC Name 2-amino-1H-pyrimidin-6-one
Formula Weight 111.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Isocytosine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 203-592-

RUO – Research Use Only