Learn More
Thermo Scientific Chemicals Isocytosine, 99%
CAS: 108-53-2 | C4H5N3O | 111.10 g/mol
Supplier: Thermo Scientific Chemicals H5422806
Description
Isocytosine (2-aminouracil) is an isomer of cytosine used in physical chemical studies involving metal complex binding, hydrogen-bonding, and tautomerism and proton transfer effects in nucleobases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsocytosine (2-aminouracil) is an isomer of cytosine used in physical chemical studies involving metal complex binding, hydrogen-bonding, and tautomerism and proton transfer effects in nucleobases.
Solubility
Soluble in DMF, DMSO, hot water, and AcOH.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
| Isocytosine | |
| C4H5N3O | |
| 5 g | |
| Soluble in DMF,DMSO,hot water,and AcOH. | |
| NC1=NC=CC(=O)N1 | |
| 111.10 | |
| CHEBI:55502 | |
| 99% |
| 108-53-2 | |
| MFCD00057557 MFCD00023256 | |
| isocytosine, 2-amino-4-hydroxypyrimidine, 2-aminopyrimidin-4-ol, 2-aminopyrimidin-4 1h-one, 2-amino-4-pyrimdinol, 2-amino-4-pyrimidone, 2-amino-4-oxypyrimidine, 2-aminouracil, 4-pyrimidinol, 2-amino, 4 1h-pyrimidinone, 2-amino | |
| XQCZBXHVTFVIFE-UHFFFAOYSA-N | |
| 2-amino-3,4-dihydropyrimidin-4-one | |
| 66950 | |
| 111.1 |
Chemical Identifiers
| 108-53-2 | |
| 111.10 | |
| XQCZBXHVTFVIFE-UHFFFAOYSA-N | |
| 66950 | |
| 2-amino-3,4-dihydropyrimidin-4-one |
| C4H5N3O | |
| MFCD00057557 MFCD00023256 | |
| isocytosine, 2-amino-4-hydroxypyrimidine, 2-aminopyrimidin-4-ol, 2-aminopyrimidin-4 1h-one, 2-amino-4-pyrimdinol, 2-amino-4-pyrimidone, 2-amino-4-oxypyrimidine, 2-aminouracil, 4-pyrimidinol, 2-amino, 4 1h-pyrimidinone, 2-amino | |
| CHEBI:55502 | |
| NC1=NC=CC(=O)N1 |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 203-592-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only