missing translation for 'onlineSavingsMsg'
Learn More

Isoconazole Nitrate 98.0+%, TCI America™

Catalog No. I08345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08345G Supplier TCI America Supplier No. I08345G

Specifications

CAS 24168-96-5
Melting Point 183°C
Color White-Yellow
Molecular Formula C18H15Cl4N3O4
Quantity 5 g
MDL Number MFCD01721647
Synonym isoconazole nitrate, adestan g100, travogyn, fazol, isoconazole; nitric acid, isoconazole nitrate jan, 1-2,4-dichloro-beta-2,6-dichlorobenzyloxy phenethyl imidazole nitrate, 1-2-2,6-dichlorobenzyl oxy-2-2,4-dichlorophenyl ethyl imidazole nitrate
InChI Key NNGQLSIGRSTLLU-UHFFFAOYNA-N
SMILES O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=C(Cl)C=CC=C2Cl)C(Cl)=C1
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid
Molecular Weight (g/mol) 479.14
PubChem CID 159968
Formula Weight 479.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoconazole Nitrate
Show More Show Less
RTECSNumber NI4772000
TSCA No