missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoconazole nitrate, 99%, Thermo Scientific™
Specifications
Specifications
| Melting Point | 181.0°C to 183.0°C |
| Infrared Spectrum | Authentic |
| Quantity | 5 g |
| Formula Weight | 479.14 |
| Percent Purity | 99% |
| Chemical Name or Material | Isoconazole nitrate |
Safety and Handling
| EINECSNumber | 246-051-4 |
RUO – Research Use Only