missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ IsoClean™ Cleanroom Front Snap Frocks, Bound Neck

Catalog No. 296361270
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packaging Type:
Bulk Packaged
Individually Packaged
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
296361270 2X-Large Individually Packaged
296361266 Small Individually Packaged
296361267 Medium Individually Packaged
296361268 Large Individually Packaged
296361269 X-Large Individually Packaged
190881125F 3X-Large Individually Packaged
190881124A Small Bulk Packaged
190881124B Medium Bulk Packaged
190881124C Large Bulk Packaged
190881124D X-Large Bulk Packaged
190881124E 2X-Large Bulk Packaged
190881124F 3X-Large Bulk Packaged
17988085 4X-Large Bulk Packaged
190881125G 4X-Large Individually Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 296361270 Supplier DuPont™ Supplier No. IC270BWH2X00300C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Clean processed versions available.

  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
  • Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
  • Frocks are ISO Class 5 (formerly Class 100) compatible
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic opening for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Adjustable snap at neck for better fit
  • Clean processed garments are individually packaged
  • Garments not clean processed are bulk packaged in double transparent poly liners

Specifications

Size 2X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Bound
Latex Free Yes
Size (English) Chest 40 to 43 in.
Size (Metric) Chest 101 to 109 cm
Sleeve Style Set-in Sleeve
Packaging Type Individually Packaged
Length (Metric) 111.76 cm
Length (English) Sleeve 38.75 in.
Length (Metric) Sleeve 98.4 cm
Description Clean Processed
Cleanroom Class Class 100 (ISO 5)
Length (English) 44 in.
Product Type Controlled Environments Frock
Seam Type Bound
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less