missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ IsoClean™ Cleanroom Front Snap Frocks, Bound Neck
Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Clean processed versions available.
Supplier: DuPont™ IC270BWHSM003000
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Frocks provide body-length jacket design with front snap closures for easy donning and doffing
- Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
- Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
- Frocks are ISO Class 5 (formerly Class 100) compatible
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Elastic opening for tighter fit at wrist
- Bound neck for lower particle shedding
- Adjustable snap at neck for better fit
- Clean processed garments are individually packaged
- Garments not clean processed are bulk packaged in double transparent poly liners
Specifications
| Controlled Environments Frock | |
| Tyvek | |
| Single-use | |
| Snaps | |
| Bound | |
| Yes | |
| 71 to 81 cm | |
| Bulk Packaged | |
| 99.06 cm | |
| 88.26 cm | |
| Class 100 (ISO 5) |
| Small | |
| White | |
| Non-sterile | |
| Elastic | |
| Bound | |
| 28 to 32 in. | |
| Set-in Sleeve | |
| 39 in. | |
| 34.75 in. | |
| Not Clean Processed |