missing translation for 'onlineSavingsMsg'
Learn More

Isobutyrophenone 98.0+%, TCI America™

Catalog No. I011325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I011325ML 25 mL
I0113500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I011325ML Supplier TCI America Supplier No. I011325ML

Chemical Identifiers

CAS 611-70-1
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008917
InChI Key BSMGLVDZZMBWQB-UHFFFAOYSA-N
Synonym isobutyrophenone, isopropyl phenyl ketone, 2-methylpropiophenone, 1-propanone, 2-methyl-1-phenyl, phenyl isopropyl ketone, isopropyl phenyl keton, alpha-methylpropiophenone, unii-8l53972ntz, .alpha.-methylpropiophenone, 2-methyl-1-phenyl-1-propanone
PubChem CID 69144
IUPAC Name 2-methyl-1-phenylpropan-1-one
SMILES CC(C)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 mL
Formula Weight 148.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyrophenone
EINECSNumber (3)-2673
TSCA Yes