missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric Acid 99.0+%, TCI America™

Catalog No. I010325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I010325ML 25 mL
I0103500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I010325ML Supplier TCI America Supplier No. I010325ML

Chemical Identifiers

CAS 79-31-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002658
InChI Key KQNPFQTWMSNSAP-UHFFFAOYSA-N
Synonym isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
PubChem CID 6590
ChEBI CHEBI:16135
IUPAC Name 2-methylpropanoic acid
SMILES CC(C)C(O)=O

Specifications

Melting Point -47°C
Boiling Point 155°C
Color Colorless
UN Number 2529
Formula Weight 88.11
Percent Purity ≥99.0% (GC)
Quantity 25 mL
Physical Form Liquid
Chemical Name or Material Isobutyric Acid
EINECSNumber (2)-0608
RTECSNumber NQ4375000
TSCA Yes