missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric acid, 99%

Catalog No. aal04038ap
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL04038AP 500 mL
AAL04038AE 100 mL
AAL040380F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL04038AP Supplier Thermo Scientific Chemicals Supplier No. L04038AP
Only null left

CAS: 79-31-2 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Isobutyric acid is used to prepare esters for flavors and perfumes. It is also used as a disinfecting agent, preservative and tanning agent. It finds applications in textile, varnish and the leather industry. Further, it is used as a lactation stimulant in dairy cattle. In addition to this, it is used in the preparation of tetramethylsuccinic acid and diisopropyl ketone.

Solubility
Miscible with water, alcohol, chloroform and ether. Slightly miscible with carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 79-31-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002658
InChI Key KQNPFQTWMSNSAP-UHFFFAOYSA-N
Synonym isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
PubChem CID 6590
ChEBI CHEBI:16135
IUPAC Name 2-methylpropanoic acid
SMILES CC(C)C(O)=O

Specifications

Melting Point -47°C
Boiling Point 153°C to 154°C
UN Number UN2529
Beilstein 635770
Merck Index 145155
Solubility Information Miscible with water,alcohol,chloroform and ether. Slightly miscible with carbon tetrachloride.
Formula Weight 88.11
Percent Purity 99%
Density 0.948
Quantity 500 mL
Flash Point 59°C (138°F)
Odor Pungent
Refractive Index 1.3928
Chemical Name or Material Isobutyric acid
Show More Show Less
Hazard Category H226-H302-H311-H314-H335
Hazard Statement GHS H Statement
H226-H302-H312
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYRIC ACID
EINECSNumber 201-195-7
RTECSNumber NQ4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only