missing translation for 'onlineSavingsMsg'
Learn More

Isobutyramide 98.0+%, TCI America™

Catalog No. p-7131214
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I010225G 25 g
I0102500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I010225G Supplier TCI America Supplier No. I010225G
Only null left

Chemical Identifiers

CAS 563-83-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00008019
InChI Key WFKAJVHLWXSISD-UHFFFAOYSA-N
Synonym isobutyramide, propanamide, 2-methyl, 2-methylpropionamide, isobutyrimidic acid, isobutylamide, isopropylformamide, dimethylacetoamide, unii-82uoe7b38z, dimethylacetamid, c-isopropylformamide
PubChem CID 68424
IUPAC Name 2-methylpropanamide
SMILES CC(C)C(=O)N

Specifications

Melting Point 128°C
Color White
Quantity 25 g
Formula Weight 87.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isobutyramide
RTECSNumber TX1475000
TSCA Yes