missing translation for 'onlineSavingsMsg'
Learn More

Isobutyltrimethoxysilane, 97%

Catalog No. aab2104836
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAB2104836 500 g
AAB2104814 25 g
AAB2104822 100 g
3 options

Catalog No. AAB2104836

Supplier: Thermo Scientific Chemicals B2104836

Only null left

CAS: 18395-30-7 | C7H18O3Si | 178.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18395-30-7
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.303
MDL Number MFCD00051589
InChI Key XYJRNCYWTVGEEG-UHFFFAOYSA-N
Synonym isobutyltrimethoxysilane, isobutyl trimethoxy silane, silane, trimethoxy 2-methylpropyl, trimethoxy 2-methylpropyl silane, dynasylan ibtmo, prosil 178, isobutyltrimethoxy silane, iso-butyl trimethoxysilane, acmc-1cg69, 2-methyl-1-propyltrimethoxysilane
PubChem CID 87622
IUPAC Name trimethoxy(2-methylpropyl)silane
SMILES CC(C)C[Si](OC)(OC)OC

Specifications

Boiling Point 154°C to 157°C
Quantity 500 g
UN Number UN1993
Beilstein 2233976
Flash Point 43°C (109°F)
Formula Weight 178.31
Percent Purity 97%
Refractive Index 1.396
Sensitivity Moisture sensitive
Density 0.93
Chemical Name or Material Isobutyltrimethoxysilane
Show More Show Less
Hazard Category H226-H315-H319-H336
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 242-272-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only