Learn More
Isobutylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
$144.50 - $144.50
Chemical Identifiers
| CAS | 5674-02-2 |
|---|---|
| Molecular Weight (g/mol) | 116.87 |
| MDL Number | MFCD00000471 |
| ChEBI | CHEBI:75272 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC209521000
|
Thermo Scientific Chemicals
209521000 |
100 mL | AcroSeal™ Glass bottle |
Each for $144.50
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5674-02-2 | |
| 116.87 | |
| MFCD00000471 | |
| CHEBI:75272 |
Specifications
| 5674-02-2,60-29-7 | |
| −040°C | |
| C4H9ClMg | |
| MFCD00000471 | |
| 0.83 | |
| Solubility in water: reacts. | |
| C1CC(=O)OC2=C1C3=CC=CC=C3C=C2 | |
| 116.87 | |
| CHEBI:75272 | |
| Isobutylmagnesium chloride, 2M solution in diethyl ether |
| 0.8300g/mL | |
| AcroSeal™ Glass bottle | |
| (CH3)2CHCH2MgCl | |
| 100 mL | |
| splitomicin, 1,2-dihydro-3h-naphtho 2,1-b pyran-3-one, 1,2-dihydro-3h-benzo f chromen-3-one, 1h-benzo f chromen-3 2h-one, chembl86537, 1,2-dihydrobenzo f chromen-3-one, splitomycin, tocris-1542, d09suo, splitomicin hplc , powder | |
| ISFPDBUKMJDAJH-UHFFFAOYSA-N | |
| 1,2-dihydrobenzo[f]chromen-3-one | |
| 5269 | |
| 116.87 |
Safety and Handling
GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.
GHS Signal Word: Danger
RUO – Research Use Only