missing translation for 'onlineSavingsMsg'
Learn More

Isobutylboronic acid, 97%

Catalog No. p-7030152
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2769803 1 g
AAH2769806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2769803 Supplier Thermo Scientific Chemicals Supplier No. H2769803
Only null left

CAS: 84110-40-7 | C4H11BO2 | 101.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84110-40-7
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00134156
InChI Key ZAZPDOYUCVFPOI-UHFFFAOYSA-N
Synonym isobutylboronic acid, isobutaneboronic acid, 2-methylpropyl boronic acid, 1-isobutaneboronic acid, 2-methylpropaneboronic acid, 2-methylprop-1-ylboronic acid, unii-khx76h7jhz, khx76h7jhz, 2-methyl-1-propylboronic acid, 2-methyl-1-propaneboronic acid
PubChem CID 2734395
IUPAC Name 2-methylpropylboronic acid
SMILES CC(C)CB(O)O

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Formula Weight 101.94
Percent Purity 97%
Chemical Name or Material Isobutylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only