missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine

Catalog No. p-4089405
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122502500 250 mL Glass bottle
AC122500010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122502500 Supplier Thermo Scientific Chemicals Supplier No. 122502500
Only null left

CAS: 78-81-9 | C4H11N | 73.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-81-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00008146
InChI Key KDSNLYIMUZNERS-UHFFFAOYSA-N
Synonym isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
PubChem CID 6558
ChEBI CHEBI:15997
IUPAC Name 2-methylpropan-1-amine
SMILES CC(C)CN

Specifications

Melting Point -85.0°C
Color Colorless to Yellow
Density 0.7300g/mL
Boiling Point 64.0°C to 71.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.397
Linear Formula (CH3)2CHCH2NH2
Quantity 250 mL
Beilstein 04, 163
Specific Gravity 0.73
Merck Index 15, 5177
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol and ether
Viscosity 0.6 mPa.s (20°C)
Formula Weight 73.14
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Isobutylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 201-145-4

RUO – Research Use Only