missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine 98.0+%, TCI America™

Catalog No. p-7131209
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
I009525ML 25 mL
I0095500ML 500 mL
2 options

Catalog No. I009525ML

Supplier: TCI America I009525ML

Only null left

Chemical Identifiers

CAS 78-81-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00008146
InChI Key KDSNLYIMUZNERS-UHFFFAOYSA-N
Synonym isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
PubChem CID 6558
ChEBI CHEBI:15997
IUPAC Name 2-methylpropan-1-amine
SMILES CC(C)CN

Specifications

Melting Point -83°C
Color Yellow
Boiling Point 66°C
Quantity 25 mL
UN Number 1214
Formula Weight 73.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutylamine
EINECSNumber (2)-0132
RTECSNumber NP9900000
TSCA Yes