missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine 98.0+%, TCI America™

Catalog No. I0095500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0095500ML 500 mL
I009525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0095500ML Supplier TCI America Supplier No. I0095500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-81-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00008146
InChI Key KDSNLYIMUZNERS-UHFFFAOYSA-N
Synonym isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
PubChem CID 6558
ChEBI CHEBI:15997
IUPAC Name 2-methylpropan-1-amine
SMILES CC(C)CN

Specifications

Melting Point -83°C
Color Yellow
Boiling Point 66°C
Quantity 500 mL
UN Number 1214
Formula Weight 73.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutylamine
EINECSNumber (2)-0132
RTECSNumber NP9900000
TSCA Yes