missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Vinyl Ether (stabilized with KOH) 99.0+%, TCI America™

Catalog No. p-7131212
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I009925ML 25 mL
I0099500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I009925ML Supplier TCI America Supplier No. I009925ML
Only null left

Chemical Identifiers

CAS 109-53-5
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008934
InChI Key OZCMOJQQLBXBKI-UHFFFAOYSA-N
Synonym isobutyl vinyl ether, vinyl isobutyl ether, propane, 1-ethenyloxy-2-methyl, ether, isobutyl vinyl, isobutoxyethene, isobutanol vinyl ether, 2-methyl-1-vinyloxypropane, lutanol lr 8500, vinoflex mo 400*, 1-ethenyloxy-2-methylpropane
PubChem CID 7992
IUPAC Name 1-ethenoxy-2-methylpropane
SMILES CC(C)COC=C

Specifications

Color Colorless
Boiling Point 83°C
Quantity 25 mL
UN Number 1304
Formula Weight 100.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Vinyl Ether (stabilized with KOH)
EINECSNumber (2)-0372
RTECSNumber KO1300000
TSCA Yes
Recommended Storage Refrigerator