missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Vinyl Ether (stabilized with KOH) 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 83°C |
| Quantity | 25 mL |
| UN Number | 1304 |
| Formula Weight | 100.16 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isobutyl Vinyl Ether (stabilized with KOH) |
Safety and Handling
| EINECSNumber | (2)-0372 |
| RTECSNumber | KO1300000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |