missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Palmitate 97.0+%, TCI America™

Catalog No. P065125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P065125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P065125ML Supplier TCI America Supplier No. P065125ML

Chemical Identifiers

CAS 110-34-9
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.538
MDL Number MFCD00059289
InChI Key OJIBJRXMHVZPLV-UHFFFAOYSA-N
Synonym Palmitic Acid Isobutyl Ester
PubChem CID 66967
IUPAC Name 2-methylpropyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(C)C

Specifications

Color Colorless
Boiling Point 207°C
Quantity 25 mL
Formula Weight 312.54
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Palmitate
EINECSNumber (2)-0798
TSCA Yes