missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Mercaptan 98.0+%, TCI America™

Catalog No. p-7132146
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M040425ML 25 mL
M0404500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M040425ML Supplier TCI America Supplier No. M040425ML
Only null left

Chemical Identifiers

CAS 513-44-0
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.184
MDL Number MFCD00004882
InChI Key BDFAOUQQXJIZDG-UHFFFAOYSA-N
Synonym isobutyl mercaptan, 2-methyl-1-propanethiol, isobutylmercaptan, isobutanethiol, 1-propanethiol, 2-methyl, isobutyl thiol, 1-isobutanethiol, unii-9h070ufp2x, 2-methyl propanethiol, iso-c4h9sh
PubChem CID 10558
IUPAC Name 2-methylpropane-1-thiol
SMILES CC(C)CS

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 mL
UN Number 2347
Formula Weight 90.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Mercaptan
EINECSNumber (2)-0464
RTECSNumber TZ7630000
TSCA Yes