missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Hexanoate 98.0+%, TCI America™

Catalog No. H011025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H011025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H011025ML Supplier TCI America Supplier No. H011025ML

Chemical Identifiers

CAS 105-79-3
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00048870
InChI Key UXUPPWPIGVTVQI-UHFFFAOYSA-N
Synonym isobutyl hexanoate, isobutyl caproate, hexanoic acid, 2-methylpropyl ester, hexanoic acid, isobutyl ester, 2-methyl-1-propyl caproate, n-caproic acid isobutyl ester, isobutyl caproate natural, unii-2a3x4w9gz0, fema no. 2202, iso-butyl n-hexanoate
PubChem CID 7775
ChEBI CHEBI:87421
IUPAC Name 2-methylpropyl hexanoate
SMILES CCCCCC(=O)OCC(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Hexanoate
EINECSNumber (2)-0782
RTECSNumber MO8389000
TSCA Yes