missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Cyanoacetate 98.0+%, TCI America™

Catalog No. p-7124734
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C100025ML 25 mL
C1000500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C100025ML Supplier TCI America Supplier No. C100025ML
Only null left

Chemical Identifiers

CAS 13361-31-4
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00044320
InChI Key HRGQEKKNLHJZGZ-UHFFFAOYSA-N
Synonym isobutyl cyanoacetate, isobutyl 2-cyanoacetate, acetic acid, cyano-, 2-methylpropyl ester, cyanoacetic acid, isobutyl ester, unii-id1wnz1d0s, id1wnz1d0s, cyanoacetic acid isobutyl ester, acetic acid, 2-cyano-, 2-methylpropyl ester, acetic acid, cyano-, isobutyl ester, ibcy
PubChem CID 25918
IUPAC Name 2-methylpropyl 2-cyanoacetate
SMILES CC(C)COC(=O)CC#N

Specifications

Color Colorless
Boiling Point 223°C
Quantity 25 mL
Formula Weight 141.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Cyanoacetate
EINECSNumber (2)-1549
RTECSNumber AG4250000
TSCA Yes