missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl chloroformate, 98%

Catalog No. p-150398
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176181000 100 g Glass bottle
AC176185000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC176181000 Supplier Thermo Scientific Chemicals Supplier No. 176181000

CAS: 543-27-1 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 543-27-1
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00000642
InChI Key YOETUEMZNOLGDB-UHFFFAOYSA-N
Synonym isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
PubChem CID 62365
IUPAC Name 2-methylpropyl carbonochloridate
SMILES CC(C)COC(=O)Cl

Specifications

Density 1.0475g/mL
Boiling Point 128.7°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. diisobutyl carbonate (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.4080
Linear Formula ClCO2CH2CH(CH3)2
Quantity 100 g
Beilstein 03, 12
Fieser 06,307
Specific Gravity 1.0475
Merck Index 15, 5184
Solubility Information Solubility in water: decomposes. Other solubilities: decomposes in alcohol,miscible with benzene,chloroform,acetone,toluene
Formula Weight 136.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl chloroformate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 208-840-1
TSCA TSCA

RUO – Research Use Only