missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Butyrate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 157°C |
| Quantity | 25 mL |
| UN Number | 3272 |
| Formula Weight | 144.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isobutyl Butyrate |
Safety and Handling
| EINECSNumber | (2)-0779 |
| RTECSNumber | ET5020000 |
| TSCA | Yes |