missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Acrylate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7117041
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A074725ML 25 mL
A0747500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A074725ML Supplier TCI America Supplier No. A074725ML
Only null left

Chemical Identifiers

CAS 106-63-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00042865
InChI Key CFVWNXQPGQOHRJ-UHFFFAOYSA-N
Synonym isobutyl acrylate, isobutyl propenoate, isobutyl 2-propenoate, 2-propenoic acid, 2-methylpropyl ester, acrylic acid, isobutyl ester, 2-methylpropyl acrylate, isobutylester kyseliny akrylove, propenoic acid, isobutyl ester, ccris 4828, hsdb 610
PubChem CID 7819
IUPAC Name 2-methylpropyl prop-2-enoate
SMILES CC(C)COC(=O)C=C

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 2527
Formula Weight 128.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0989
RTECSNumber AT2100000
TSCA Yes