missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Acetate 99.0+%, TCI America™

Catalog No. A003425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A003425ML 25 mL
A0034500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A003425ML Supplier TCI America Supplier No. A003425ML

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008932
InChI Key GJRQTCIYDGXPES-UHFFFAOYSA-N
Synonym isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
PubChem CID 8038
ChEBI CHEBI:50569
IUPAC Name 2-methylpropyl acetate
SMILES CC(C)COC(C)=O

Specifications

Color Colorless
Boiling Point 117°C
Quantity 25 mL
UN Number 1213
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Acetate
EINECSNumber (2)-0731
RTECSNumber AI4025000
TSCA Yes