missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl Methacrylate (stabilized with MEHQ) 85.0+%, TCI America™

Catalog No. I061725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I061725G 25 g
I0617500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I061725G Supplier TCI America Supplier No. I061725G

Chemical Identifiers

CAS 7534-94-3
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00081070
InChI Key IAXXETNIOYFMLW-UHFFFAOYNA-N
Synonym ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate
PubChem CID 71311141
IUPAC Name 1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C

Specifications

Color Yellow
Boiling Point 129°C
Quantity 25 g
Formula Weight 222.33
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobornyl Methacrylate (stabilized with MEHQ)
EINECSNumber (4)-1492
TSCA Yes