missing translation for 'onlineSavingsMsg'
Learn More

Isoamylamine 98.0+%, TCI America™

Catalog No. p-7131203
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I008225ML 25 mL
I0082100ML 100 mL
I0082500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I008225ML Supplier TCI America Supplier No. I008225ML

Chemical Identifiers

CAS 107-85-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008203
InChI Key BMFVGAAISNGQNM-UHFFFAOYSA-N
Synonym isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine
PubChem CID 7894
ChEBI CHEBI:43689
IUPAC Name 3-methylbutan-1-amine
SMILES CC(C)CCN

Specifications

Color Yellow
Boiling Point 97°C
Quantity 25 mL
UN Number 1106
Formula Weight 87.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Isoamylamine
EINECSNumber (2)-0133
TSCA Yes