missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Propionate 98.0+%, TCI America™

Catalog No. I067725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I067725G 25 g
I0677500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I067725G Supplier TCI America Supplier No. I067725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-68-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00048611
InChI Key XAOGXQMKWQFZEM-UHFFFAOYSA-N
Synonym isoamyl propionate, isoamyl propanoate, isopentyl propanoate, 3-methylbutyl propionate, isopentyl propionate, isopentyl alcohol, propionate, 1-butanol, 3-methyl-, propanoate, iso-pentyl propionate, propionic acid, isopentyl ester, 3-methyl-1-butyl propanoate
PubChem CID 7772
ChEBI CHEBI:87419
IUPAC Name 3-methylbutyl propanoate
SMILES CCC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 161°C
Quantity 25 g
UN Number 3272
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Propionate
EINECSNumber (2)-0774
RTECSNumber NT0190000
TSCA Yes