missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Nitrite 95.0+%, TCI America™

Catalog No. p-7131206
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I008925ML 25 mL
I0089100ML 100 mL
I0089500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I008925ML Supplier TCI America Supplier No. I008925ML

Chemical Identifiers

CAS 110-46-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00002057
InChI Key OWFXIOWLTKNBAP-UHFFFAOYSA-N
Synonym isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i
PubChem CID 8053
ChEBI CHEBI:2691
IUPAC Name 3-methylbutyl nitrite
SMILES CC(C)CCON=O

Specifications

Color Yellow
Boiling Point 99°C
Quantity 25 mL
UN Number 1113
Formula Weight 117.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Nitrite
EINECSNumber (2)-2796
RTECSNumber NT0187500
TSCA Yes
Recommended Storage Refrigerator