missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Isovalerate 98.0+%, TCI America™

$34.66 - $202.15

Chemical Identifiers

CAS 659-70-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00048369
InChI Key XINCECQTMHSORG-UHFFFAOYSA-N
Synonym Isovaleric Acid Isoamyl Ester, Isopentyl Isovalerate, Isovaleric Acid Isopentyl Ester
PubChem CID 12613
ChEBI CHEBI:31727
IUPAC Name 3-methylbutyl 3-methylbutanoate
SMILES CC(C)CCOC(=O)CC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I019625ML
View Documents
TCI America
I019625ML
25 mL
Each for $34.66
Only null left
 
I0196500ML
View Documents
TCI America
I0196500ML
500 mL
Each for $202.15
Only null left
 
Description

Description

Specifications

Chemical Identifiers

659-70-1
172.268
XINCECQTMHSORG-UHFFFAOYSA-N
12613
3-methylbutyl 3-methylbutanoate
C10H20O2
MFCD00048369
Isovaleric Acid Isoamyl Ester, Isopentyl Isovalerate, Isovaleric Acid Isopentyl Ester
CHEBI:31727
CC(C)CCOC(=O)CC(C)C

Specifications

659-70-1
194°C
MFCD00048369
Isovaleric Acid Isoamyl Ester, Isopentyl Isovalerate, Isovaleric Acid Isopentyl Ester
CC(C)CCOC(=O)CC(C)C
172.268
CHEBI:31727
≥98.0% (GC)
Isoamyl Isovalerate
Colorless
C10H20O2
25 mL
XINCECQTMHSORG-UHFFFAOYSA-N
3-methylbutyl 3-methylbutanoate
12613
172.27
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0776

RTECSNumber : NY1508000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'