missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Isovalerate 98.0+%, TCI America™

Catalog No. I0196500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0196500ML 500 mL
I019625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0196500ML Supplier TCI America Supplier No. I0196500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 659-70-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00048369
InChI Key XINCECQTMHSORG-UHFFFAOYSA-N
Synonym Isovaleric Acid Isoamyl Ester, Isopentyl Isovalerate, Isovaleric Acid Isopentyl Ester
PubChem CID 12613
ChEBI CHEBI:31727
IUPAC Name 3-methylbutyl 3-methylbutanoate
SMILES CC(C)CCOC(=O)CC(C)C

Specifications

Color Colorless
Boiling Point 194°C
Quantity 500 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Isovalerate
EINECSNumber (2)-0776
RTECSNumber NY1508000
TSCA Yes