missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Butyrate 98.0+%, TCI America™

Catalog No. B076025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B076025ML 25 mL
B0760500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B076025ML Supplier TCI America Supplier No. B076025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-27-4
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00044888
InChI Key PQLMXFQTAMDXIZ-UHFFFAOYSA-N
Synonym isoamyl butyrate, isopentyl butyrate, isopentyl butanoate, isoamyl butanoate, butanoic acid, 3-methylbutyl ester, 3-methylbutyl butyrate, butyric acid, isopentyl ester, isoamyl-n-butyrate, isopentyl alcohol, butyrate, isoamyl butylate
PubChem CID 7795
ChEBI CHEBI:87422
IUPAC Name 3-methylbutyl butanoate
SMILES CCCC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 179°C
Quantity 25 mL
UN Number 2620
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Butyrate
EINECSNumber (2)-0779
RTECSNumber ET5034000
TSCA Yes